missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 11 Recombinant Rabbit Monoclonal Antibody (23GB5090), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552786
This item is not returnable.
View return policy
Description
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.
Specifications
| Claudin 11 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Cldn11 | |
| claudin 11; claudin 11 (oligodendrocyte transmembrane protein); Claudin11; claudin-11; Cldn11; hypothetical protein LOC508268; oligodendrocyte transmembrane protein; Oligodendrocyte-specific protein; OSP; OTM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5010 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 23GB5090 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75508 | |
| Cldn11 | |
| A synthesized peptide derived from human Oligodendrocyte Specific Protein (25-80AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction