missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99148-50ul
This item is not returnable.
View return policy
Description
Claudin 10 Polyclonal antibody specifically detects Claudin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Claudin 10 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| claudin 10, claudin-10, CPETRL3, OSP-L, OSP-like protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Claudin 10. | |
| 50 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9071 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction