missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£190.00 - £255.00
Specifications
| Antigen | Claudin 10 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645685
|
Novus Biologicals
NBP2-99148-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698945
|
Novus Biologicals
NBP2-99148-50ul |
50 μg |
£190.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin 10 Polyclonal antibody specifically detects Claudin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Claudin 10 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 9071 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| claudin 10, claudin-10, CPETRL3, OSP-L, OSP-like protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Claudin 10. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts