missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
£150.00
Specifications
| Antigen | Claudin-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
Claudin-1 Polyclonal specifically detects Claudin-1 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.Specifications
| Claudin-1 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Extracellular Matrix | |
| PBS with 0.02% Sodium Azide | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| CLDN1 | |
| IgG | |
| Affinity Purified | |
| 23 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| O95832 | |
| 9076 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human CLDN1. The immunogen is located within the last 50 amino acids of CLDN1. Amino Acid Squence: PRKTTSYPTPRPYPKPAPS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title