missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NBP1-77036-0.025mg
This item is not returnable.
View return policy
Description
Claudin-1 Polyclonal specifically detects Claudin-1 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunocytochemistry.
Specifications
| Claudin-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
| Rabbit | |
| 23 kDa | |
| 0.025 mg | |
| Extracellular Matrix | |
| 9076 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 5-20 μg/mL | |
| O95832 | |
| CLDN1 | |
| Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human CLDN1. The immunogen is located within the last 50 amino acids of CLDN1. Amino Acid Squence: PRKTTSYPTPRPYPKPAPS | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction