missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M1 Weights
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
£117.66 - £213.06
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
12001613
|
Zwiebel™
PBI23717 |
±500 mg |
£165.36
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12011613
|
Zwiebel™
PBI23718 |
±1000 mg |
£213.06
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11794724
|
Zwiebel™
PBI23716 |
±250 mg |
£117.66
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- Class M1 cast iron calibration weight
- In compliance with OIML R111
- Supplied individually
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |