missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M1 Weights
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weights are ideal for the verification or calibration of class M2 weights.
Brand: Zwiebel™ PBI23716
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Class M1 cast iron calibration weight
- In compliance with OIML R111
- Supplied individually
Specifications
±250 mg | |
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |