missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class M1 Weight Boxes
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weight Boxes are ideal for the verification or calibration of class M₂ weights.
£110.24 - £232.14
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11840892
|
Zwiebel™
PBI56815 |
1000 g |
£160.06
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
11850902
|
Zwiebel™
PBI5681Y |
2000 g |
£232.14
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
11870872
|
Zwiebel™
PBI5679Y |
500 g |
£110.24
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
Description
- Class M1 calibration weight
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |