missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Weight Boxes

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Weight Boxes are ideal for the verification or calibration of class M₂ weights.

Brand:  Zwiebel™ PBI5679Y

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11870872

  • £110.24 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

  • Class M1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications

Specifications

500 g
±25 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers