Learn More
Zwiebel™ Class F2 Box Weights
£45.16 - £285.14
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11840912
|
Zwiebel™
PBI56829 |
10 g | ±0.6 mg |
£58.30
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
11870912
|
Zwiebel™
PBI5682C |
100 g | ±1.6 mg |
£70.12
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
11800922
|
Zwiebel™
PBI5682F |
1000 g | ±15 mg |
£212.00
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11810912
|
Zwiebel™
PBI56825 |
1 g | ±0.3 mg |
£45.16
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11810922
|
Zwiebel™
PBI5682H |
2000 g | ±30 mg |
£285.14
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
11880912
|
Zwiebel™
PBI5682D |
200 g | ±3 mg |
£90.79
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
11820912
|
Zwiebel™
PBI56827 |
2 g | ±0.4 mg |
£47.01
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11890912
|
Zwiebel™
PBI5682E |
500 g | ±7.5 mg |
£169.60
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
Specifications
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |