Learn More
Zwiebel™ Class F2 Box Weights
Brand:  Zwiebel™ PBI5682C
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11870912
  • £70.12 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Specifications
100 g
±1.6 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II