Learn More
Zwiebel™ Class E2 Box Weight
£59.25 - £267.12
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11780623
|
Zwiebel™
PBI2302B |
10 mg | ±0.008 mg |
£59.25
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11720673
|
Zwiebel™
PBI23063 |
10 g | ±0.06 mg |
£152.64
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11730673
|
Zwiebel™
PBI23064 |
20 g | ±0.08 mg |
£153.70
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
11760673
|
Zwiebel™
PBI23067 |
200 g | ±0.3 mg |
£267.12
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
11700673
|
Zwiebel™
PBI23061 |
2 g | ±0.04 mg |
£134.62
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11790663
|
Zwiebel™
PBI23060 |
1 g | ±0.03 mg |
£124.02
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11750623
|
Zwiebel™
PBI2301Z |
2 mg | ±0.006 mg |
£59.25
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11730633
|
Zwiebel™
PBI2302H |
500 mg | ±0.025 mg |
£59.25
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11710673
|
Zwiebel™
PBI23062 |
5 g | ±0.05 mg |
£139.92
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
11740673
|
Zwiebel™
PBI23065 |
50 g | ±0.1 mg |
£179.14
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
Specifications
For use with Weighing Instruments of Special Accuracy Class I |