Learn More
Zwiebel™ Class E2 Box Weight
Brand:  Zwiebel™ PBI2301Z
Code : A0
Additional Details : Weight : 0.01800kg
 View more versions of this product
Product Code. 11750623
  • £59.25 / Each
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Specifications
2 mg
±0.006 mg
For use with Weighing Instruments of Special Accuracy Class I