Learn More
Zwiebel™ Class E2 Box Weight
Brand: Zwiebel™ PBI23060
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
1 g | |
±0.03 mg | |
For use with Weighing Instruments of Special Accuracy Class I |