missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cl-C6-PEG4-O-CH2COOH

Product Code. 16465476
Encompass_Preferred
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16465476 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16465476 Supplier MedChemExpress Supplier No. HY112496

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cl-C6-PEG4-O-CH2COOH (PROTAC Linker 4) is a PEG-based PROTAC linker can be used in the synthesis of chloroalkane-containing PROTACs (HaloPROTACs).

Specifications

Molecular Weight (g/mol) 370.87 g/mol
CAS 1799506-30-1
Content And Storage Pure form -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Quantity 100 mg
Synonym PROTAC Linker 4
Solubility Ethanol : 100 mg/mL (269.64 mM; Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.