missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKII alpha prime polypeptide Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56402
This item is not returnable.
View return policy
Description
CKII alpha prime polypeptide Polyclonal specifically detects CKII alpha prime polypeptide in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| CKII alpha prime polypeptide | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| casein kinase 2, alpha prime polypeptide, casein kinase II subunit alpha', CK II alpha', CK2A2, CSNK2A1, EC 2.7.11, EC 2.7.11.1, FLJ43934 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: alpha'. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunoprecipitation | |
| P19784 | |
| CSNK2A2 | |
| Synthetic peptides corresponding to CSNK2A2(casein kinase 2, alpha prime polypeptide) The peptide sequence was selected from the C terminal of CSNK2A2. Peptide sequence LLDKLLRYDHQQRLTAKEAMEHPYFYPVVKEQSQPCADNAVLSSGLTAAR. | |
| 100 μL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 1459 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction