missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin Modified DNA Antibody (CP9/19) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£176.00 - £333.00
Specifications
| Antigen | Cisplatin Modified DNA |
|---|---|
| Clone | CP9/19 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot 1:100 - 1:2000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654977
|
Novus Biologicals
NBP2-50165-0.025ml |
0.025 mL |
£176.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18611548
|
Novus Biologicals
NBP2-50165 |
0.1 mL |
£333.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot BlotSpecifications
| Cisplatin Modified DNA | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rat | |
| DNA Repair, Epigenetics | |
| PBS | |
| Cisplatin modified native DNA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CP9/19 | |
| Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| ICR4 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title