missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cisplatin Modified DNA Antibody (CP9/19) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-50165-0.025ml
This item is not returnable.
View return policy
Description
Cisplatin Modified DNA Monoclonal antibody specifically detects Cisplatin Modified DNA in Nonspecies specific samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot
Specifications
| Cisplatin Modified DNA | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot 1:100 - 1:2000 | |
| ICR4 | |
| Cisplatin modified native DNA | |
| 0.025 mL | |
| DNA Repair, Epigenetics | |
| Non-species specific | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CP9/19 | |
| Unconjugated | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction