missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-endo-2,3-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15440178
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440178 5 g 5g
15450178 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440178 Supplier Thermo Scientific Alfa Aesar Supplier No. H53403.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3853-88-1 | Molecular Formula: C9H10O4 | Molecular Weight: 182.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3853-88-1
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00003735
InChI Key NIDNOXCRFUCAKQ-ZUMNNYEFSA-N
Synonym cis-5-norbornene-endo-2,3-dicarboxylic acid
PubChem CID 122176685
IUPAC Name (2S,3R,4S)-bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
SMILES C1C2C=CC1C(C2C(=O)O)C(=O)O

Specifications

Melting Point 175°C (decomposition)
Quantity 5 g
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.