accessibility menu, dialog, popup

UserName

CAS RN 3853-88-1

IUPAC Name: bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Synonyms: bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: NIDNOXCRFUCAKQ-UHFFFAOYNA-N
SMILES: OC(=O)C1C2CC(C=C2)C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results