missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Boc-amino)cyclohexanemethanol, 97%

Product Code. 15499218
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499218 1 g 1g
15409228 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499218 Supplier Thermo Scientific Alfa Aesar Supplier No. H62744.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 920966-16-1 | C12H23NO3 | 229.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920966-16-1
Molecular Formula C12H23NO3
Molecular Weight (g/mol) 229.32
MDL Number MFCD03844608
InChI Key LBJSEPNOVVUVJA-ZJUUUORDSA-N
Synonym tert-Butyl cis-(3-hydroxymethyl)cyclohexylcarbamate; cis-1-(Boc-amino)-3-(hydroxymethyl)cyclohexane
PubChem CID 83823767
IUPAC Name tert-butyl N-[(1S,3R)-3-(hydroxymethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)CO

Specifications

Quantity 1 g
Formula Weight 229.31
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclohexanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.