CAS RN 920966-16-1
CAS RN 920966-16-1
IUPAC Name:
tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Synonyms:
tert-butyl N-[3-(hydroxymethyl)cyclohexyl]carbamate
Molecular Weight (g/mol):
229.32
Molecular Formula:
C12H23NO3
InChi Key:
LBJSEPNOVVUVJA-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)NC1CCCC(CO)C1
1
–
1
of
1
results
1
cis-3-(Boc-amino)cyclohexanemethanol, 97%
CAS: 920966-16-1 Molecular Formula: C12H23NO3 Molecular Weight (g/mol): 229.32 MDL Number: MFCD03844608 InChI Key: LBJSEPNOVVUVJA-ZJUUUORDSA-N Synonym: tert-Butyl cis-(3-hydroxymethyl)cyclohexylcarbamate; cis-1-(Boc-amino)-3-(hydroxymethyl)cyclohexane PubChem CID: 83823767 IUPAC Name: tert-butyl N-[(1S,3R)-3-(hydroxymethyl)cyclohexyl]carbamate SMILES: CC(C)(C)OC(=O)NC1CCCC(C1)CO