missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIRBP Recombinant Rabbit Monoclonal Antibody (23GB7980), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | CIRBP |
|---|---|
| Clone | 23GB7980 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089922
![]() |
Invitrogen™
MA553306 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088013
|
Invitrogen™
MA553307 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CIRBP Recombinant Monoclonal antibody specifically detects CIRBP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CIRBP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q14011 | |
| 1153 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB7980 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39 | |
| A synthesized peptide derived from CIRBP (1-172AA). | |
| Primary | |
| -20°C | |
| CIRBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title