missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CIRBP Recombinant Rabbit Monoclonal Antibody (23GB7980), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553307
This item is not returnable.
View return policy
Description
CIRBP is cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. It acts as a translational activator and seems to play an essential role in cold-induced suppression of cell proliferation. It binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. This protein acts as a translational repressor and promotes assembly of stress granules (SGs), when overexpressed.
Specifications
| CIRBP | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CIRBP | |
| A18 hnRNP; A18HNRNP; CIRBP; CIRP; cold inducible RNA binding protein; cold inducible RNA-binding protein; cold-inducible RNA-binding protein; glycine-rich RNA binding protein; glycine-rich RNA-binding protein CIRP; R74941; testicular tissue protein Li 39 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1153 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB7980 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14011 | |
| CIRBP | |
| A synthesized peptide derived from CIRBP (1-172AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction