missing translation for 'onlineSavingsMsg'
Learn More

Cinoxate, TRC

Product Code. 30352425
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352425 2.5 g 2.5g
30367075 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352425 Supplier Toronto Research Chemicals Supplier No. TRCC4758002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-28-9
Molecular Formula C14 H18 O4
Molecular Weight (g/mol) 250.29
Synonym 3-(4-Methoxyphenyl)-2-propenoic Acid 2-Ethoxyethyl Ester, p-Methoxycinnamic Acid 2-Ehoxyethyl Ester, 2-Ethoxyehanol p-Methoxycinnamate, 2-Ethoxyethyl 4-Methoxycinnamate, 2-Ethoxyethyl p-Methoxycinnamate, Giv Tan F, Give-Tan, Phiasol, Sundare
IUPAC Name 2-ethoxyethyl (E)-3-(4-methoxyphenyl)prop-2-enoate
SMILES CCOCCOC(=O)\C=C\c1ccc(OC)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H18O4/c1-3-17-10-11-18-14(15)9-6-12-4-7-13(16-2)8-5-12/h4-9H,3,10-11H2,1-2H3/b9-6+
Formula Weight 250.1205
Percent Purity >95%
Chemical Name or Material Cinoxate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.