CAS RN 104-28-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cinoxate, TRC
CAS: 104-28-9 Molecular Formula: C14 H18 O4 Molecular Weight (g/mol): 250.29 Synonym: 3-(4-Methoxyphenyl)-2-propenoic Acid 2-Ethoxyethyl Ester,p-Methoxycinnamic Acid 2-Ehoxyethyl Ester,2-Ethoxyehanol p-Methoxycinnamate,2-Ethoxyethyl 4-Methoxycinnamate,2-Ethoxyethyl p-Methoxycinnamate,Giv Tan F,Give-Tan,Phiasol,Sundare IUPAC Name: 2-ethoxyethyl (E)-3-(4-methoxyphenyl)prop-2-enoate SMILES: CCOCCOC(=O)\C=C\c1ccc(OC)cc1