missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£150.00
Specifications
| Antigen | CIDEA |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
CIDEA Polyclonal specifically detects CIDEA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| CIDEA | |
| Western Blot 0.5 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
| CIDEA | |
| IgG | |
| Affinity Purified | |
| It has no cross activity to CIDE-B. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAC34985 | |
| 1149 | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of mouse CIDE-A. The immunogen is located within the last 50 amino acids of CIDE-A. Amino Acid Squence: GDTEEQPSPKPSTKG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 25 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title