missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIDEA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-76950-0.025mg
This item is not returnable.
View return policy
Description
CIDEA Polyclonal specifically detects CIDEA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| CIDEA | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| cell death activator CIDE-A, cell death-inducing DFFA-like effector aCIDE-A | |
| Rabbit | |
| 25 kDa | |
| 0.025 mg | |
| Apoptosis | |
| 1149 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| AAC34985 | |
| CIDEA | |
| Antibody was raised against an 18 amino acid peptide near the carboxy terminus of mouse CIDE-A. The immunogen is located within the last 50 amino acids of CIDE-A. Amino Acid Squence: GDTEEQPSPKPSTKG | |
| Affinity Purified | |
| RUO | |
| Primary | |
| It has no cross activity to CIDE-B. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction