missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chymotrypsin Monoclonal Antibody (CHYMO10-104.10), Invitrogen™
Mouse Monoclonal Antibody
£367.00 - £916.00
Specifications
| Antigen | Chymotrypsin |
|---|---|
| Clone | CHYMO10-104.10 |
| Concentration | 2.0 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17870346
|
Invitrogen™
MA5-46156 |
100 μg |
£367.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11548912
|
Invitrogen™
MA110624 |
500 μg |
£442.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17820486
|
Invitrogen™
MA5-46015 |
1 mg |
£916.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10624 detects Chymotrypsin from human samples. MA1-10624 has been successfully used in ELISA and Western blot applications. The MA1-10624 immunogen is purified human pancreatic Chymotrypsin.
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.Specifications
| Chymotrypsin | |
| 2.0 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| 1810044E12Rik; caldecrin; chymotrypsin C; chymotrypsin C (caldecrin); chymotrypsin-C; CLCR; Ctrc; ELA4; elastase 4; elastase IV; preprocaldecrin; serum calcium decreasing factor; serum calcium-decreasing factor | |
| CTRC | |
| IgG3 | |
| Protein A | |
| Antibody |
| CHYMO10-104.10 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99895 | |
| 11330 | |
| Purified human pancreatic Chymotrypsin. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| CTRC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title