missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chymotrypsin Monoclonal Antibody (CHYMO10-104.10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110624
This item is not returnable.
View return policy
Description
MA1-10624 detects Chymotrypsin from human samples. MA1-10624 has been successfully used in ELISA and Western blot applications. The MA1-10624 immunogen is purified human pancreatic Chymotrypsin.
This gene encodes a member of the peptidase S1 family. The encoded protein is a serum calcium-decreasing factor that has chymotrypsin-like protease activity. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
| Chymotrypsin | |
| Monoclonal | |
| 2.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q99895 | |
| CTRC | |
| Purified human pancreatic Chymotrypsin. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG3 |
| ELISA | |
| CHYMO10-104.10 | |
| Unconjugated | |
| CTRC | |
| 1810044E12Rik; caldecrin; chymotrypsin C; chymotrypsin C (caldecrin); chymotrypsin-C; CLCR; Ctrc; ELA4; elastase 4; elastase IV; preprocaldecrin; serum calcium decreasing factor; serum calcium-decreasing factor | |
| Mouse | |
| Protein A | |
| RUO | |
| 11330 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction