missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymotrypsin C/CTRC Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-58035
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Chymotrypsin C/CTRC Polyclonal specifically detects Chymotrypsin C/CTRC in Human samples. It is validated for Western Blot.
Especificaciones
| Chymotrypsin C/CTRC | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Caldecrin, chymotrypsin C (caldecrin), chymotrypsin-C, CLCRserum calcium decreasing factor, EC 3.4.21, EC 3.4.21.2, ELA4, elastase 4, elastase IV | |
| Rabbit | |
| 27 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Canine: 85%; Mouse: 85%; Bovine: 78%; Rat: 78%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q99895 | |
| CTRC | |
| Synthetic peptides corresponding to CTRC(chymotrypsin C (caldecrin)) The peptide sequence was selected from the N terminal of CTRC. Peptide sequence LGITVLAALLACASSCGVPSFPPNLSARVVGGEDARPHSWPWQISLQYLK. | |
| Affinity purified | |
| RUO | |
| 11330 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido