missing translation for 'onlineSavingsMsg'
Learn More

Chymase/CMA1/Mast Cell Chymase Antibody (CC1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

£141.00 - £589.00

Specifications

Antigen Chymase/CMA1/Mast Cell Chymase
Clone CC1
Classification Monoclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18288104
View Documents
Novus Biologicals
NB100-693-0.1ML
0.1 mL
£141.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18714973
View Documents
Novus Biologicals
NB100-693
0.5 mL
£589.00
0.50mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Chymase/CMA1/Mast Cell Chymase
Monoclonal
Purified
RUO
Human
Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891
CMA1
IgG1
Protein A or G purified
This reacts with mast cells distributed in skin, synovium, lung and heart.
CC1
Unconjugated
Mouse
Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Mast Cell Markers
P23946
1215
BALB/C mice were injected with a purified human skin chymase.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.