missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymase/CMA1/Mast Cell Chymase Antibody (CC1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-693-0.1ML
117.30 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Chymase/CMA1/Mast Cell Chymase | |
| Monoclonal | |
| Unconjugated | |
| Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCT1, MGC119890, MGC119891 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This reacts with mast cells distributed in skin, synovium, lung and heart. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunocytochemistry | |
| CC1 | |
| Immunohistochemistry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:25-1:100 | |
| P23946 | |
| CMA1 | |
| BALB/C mice were injected with a purified human skin chymase. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Mast Cell Markers | |
| 1215 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction