missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | CHST14 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
CHST14 Polyclonal specifically detects CHST14 in Human samples. It is validated for Western Blot.Specifications
| CHST14 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| 113189 | |
| IgG | |
| 35 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ATCS, carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14, D4ST-1, D4ST1carbohydrate sulfotransferase 14, dermatan 4 sulfotransferase 1, Dermatan 4-sulfotransferase 1, EC 2.8.2.-, HD4ST, hD4ST1, HNK1ST | |
| CHST14 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title