missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55174
This item is not returnable.
View return policy
Description
CHST14 Polyclonal specifically detects CHST14 in Human samples. It is validated for Western Blot.
Specifications
| CHST14 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CHST14 | |
| Synthetic peptides corresponding to CHST14(carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14) The peptide sequence was selected from the middle region of CHST14. Peptide sequence REYQQRYGAEIVRRYRAGAGPSPAGDDVTFPEFLRYLVDEDPERMNEHWM The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Guinea pig: 86%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| ATCS, carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 14, D4ST-1, D4ST1carbohydrate sulfotransferase 14, dermatan 4 sulfotransferase 1, Dermatan 4-sulfotransferase 1, EC 2.8.2.-, HD4ST, hD4ST1, HNK1ST | |
| Rabbit | |
| 35 kDa | |
| 100 μL | |
| Lipid and Metabolism | |
| 113189 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction