missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHST13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-62294
This item is not returnable.
View return policy
Description
CHST13 Polyclonal specifically detects CHST13 in Human samples. It is validated for Western Blot.
Specifications
| CHST13 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C4ST-3, C4ST3MGC119279, carbohydrate (chondroitin 4) sulfotransferase 13, carbohydrate sulfotransferase 13, Chondroitin 4-O-sulfotransferase 3, Chondroitin 4-sulfotransferase 3, EC 2.8.2.5, MGC119278, MGC119281 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Lipid and Metabolism | |
| 166012 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8NET6 | |
| CHST13 | |
| Synthetic peptides corresponding to CHST13(carbohydrate (chondroitin 4) sulfotransferase 13) The peptide sequence was selected from the C terminal of CHST13. Peptide sequence CHPCRLRYDVVGKFETLAEDAAFVLGLAGASDLSFPGPPRPRGAAASRDL. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Pig: 77%; Guinea pig: 77%. | |
| Human, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction