Learn More
Proteintech ChromoTek V5-Trap™ Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- V5-Trap™ Agarose is an affinity reagent for IP and purification of V5-tagged proteins
- Consists of an anti-V5 Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |