Learn More
Proteintech ChromoTek TurboGFP-Trap Agarose Beads Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- TurboGFP-Trap Agarose is an affinity bead for immunoprecipitation (IP) of TurboGFP fusion proteins
- Comprises an anti-TurboGFP VHH/ Nanobody coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |