Learn More
Proteintech ChromoTek RFP-Trap™ Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- RFP-Trap™ Agarose is an affinity resin for IP of RFP-fusion proteins
- Consists of an RFP Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |