Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins
- Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |