Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins.
Brand: Proteintech NTA-400
Additional Details : Weight : 0.00480kg
Description
- Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Upon receipt store at 4°C. Do not freeze. | |
mNeonGreen, Branchiostoma lanceolatum | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose | |
400 Reactions |