Learn More
Proteintech ChromoTek mNeonGreen-Trap Agarose Beads
mNeonGreen-Trap Agarose is an affinity reagent for IP of mNeonGreen-fusion proteins.
Brand:  Proteintech NTA-10
Additional Details : Weight : 0.00240kg
 View more versions of this product
Product Code. 17303393
  • £208.00 / 10 reactions
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
  • Consists of a mNeonGreen Nanobody/ VHH coupled to agarose beads
Specifications
Specifications
Upon receipt store at 4°C. Do not freeze.
mNeonGreen, Branchiostoma lanceolatum
Alpaca
IP, CoIP, ChIP, RIP
Agarose
10 Reactions