Learn More
Proteintech ChromoTek MBP-Trap Agarose Beads Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- The ChromoTek MBP-Trap Agarose are affinity beads for IP of MBP-fusion proteins
- They comprise MBP Nanobodies/ VHHs coupled to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |