Learn More
Proteintech ChromoTek iST RFP-Trap Kit
Product Code. 17373443
Click to view available options
Quantity:
8 Reactions
Unit Size:
8 reactions
Product Code. 17373443
Brand: Proteintech rtakiST8
Kit for AP-MS sample preparation of RFP-fusion proteins. Includes 8 reactions plus 2 controls.
  • Kit for IP and Co-IP of RFP- and mCherry fusion proteins & sample preparation for mass spectrometry (MS)
Specifications
Quantity 8 Reactions
Sufficient For 8 Reactions