missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Trap™ Agarose

Product Code. 17373343
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
This item is not returnable. View return policy

Product Code. 17373343

Brand: Proteintech gta10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GFP-Trap™ Agarose is an affinity resin for IP of GFP-fusion proteins.

  • Consists of a GFP Nanobody/ VHH coupled to agarose beads

Specifications

Binding Capacity 10 μL slurry bind about 12 μg of recombinant GFP
Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP
Label or Dye Agarose
Species Alpaca
Sufficient For 10 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.