missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin B Recombinant Rabbit Monoclonal Antibody (23GB5075), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Chromogranin B |
|---|---|
| Clone | 23GB5075 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088140
|
Invitrogen™
MA552780 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087841
|
Invitrogen™
MA552781 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromogranin B Recombinant Monoclonal antibody specifically detects Chromogranin B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Chromogranin B | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O35314, P05060, P16014 | |
| 1114, 12653, 24259 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5075 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI | |
| A synthesized peptide derived from human Chromogranin B (600-677AA). | |
| Primary | |
| -20°C | |
| Chgb |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title