missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chromogranin B Recombinant Rabbit Monoclonal Antibody (23GB5075), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552781
This item is not returnable.
View return policy
Description
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.
Specifications
| Chromogranin B | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Chgb | |
| CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1114, 12653, 24259 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5075 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35314, P05060, P16014 | |
| Chgb | |
| A synthesized peptide derived from human Chromogranin B (600-677AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction