missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Rabbit, Clone: CHGA/1773R, enQuireBio™
Rabbit Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Chromogranin A |
---|---|
Clone | CHGA/1773R |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15949797
|
EnquireBio
1113-RBM13-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939797
|
EnquireBio
1113-RBM13-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifica
Chromogranin A | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
1113 | |
Recombinant human full-length Chromogranin A protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
68-75kDa |
CHGA/1773R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001266 | |
CHGA | |
IgG κ | |
Protein A/G Purified | |
Finely granular cytoplasmic | |
PC12 cells. Adrenal gland, bowel, parathyroid, pancreas, or pheochromocytoma. |