Learn More
Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.
£95.15 - £335.00
Specifications
| Antigen | Chromogranin A Ab-1 |
|---|---|
| Clone | LK2H10 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Form | Concentration | Price | Quantity & Availability | ||
|
12651007
|
Epredia
MS-324-B0 |
100 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12661007
|
Epredia
MS-324-B1 |
500 μL | Biotin | Concentrated | 200μg/mL | ||||
|
12681007
|
Epredia
MS-324-P0 |
100 μL | Unconjugated | Concentrated | 200μg/mL | ||||
|
12601017
|
Epredia
MS-324-P1ABX |
100 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12611017
|
Epredia
MS-324-PABX |
200 μL | Unconjugated | Concentrated | 1mg/mL | ||||
|
12691007
|
Epredia
MS-324-P1 |
500 μL | Unconjugated | Concentrated | 200μg/mL |
£230.00 £215.65 / 500µL Save £14.35 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
|
12671007
|
Epredia
MS-324-P |
1 mL | Unconjugated | Concentrated | 200μg/mL |
£325.00 £305.23 / 1mL Save £19.77 6% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
|
12641017
|
Epredia
MS-324-R7 |
7 mL | Unconjugated | Prediluted | Prediluted | ||||
Description
Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.Host Species: Mouse
Clone: LK2H10
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: PC-12 cells, Pancreas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
| Chromogranin A Ab-1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Mouse | |
| Oncogenesis | |
| Human pheochromocytoma | |
| Primary |
| LK2H10 | |
| Monoclonal | |
| IVD | |
| Human, Non-human Primate, Pig, Rabbit, Rat | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.