missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Chromogranin A Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-324-B1

 View more versions of this product

Product Code. 12661007

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Chromogranin A (a protein of 439-amino acids which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: LK2H10

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Pig, Rat and Rabbit. Others not known

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: PC-12 cells, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chromogranin A Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Oncogenesis
Human, Non-human Primate, Pig, Rabbit, Rat
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
LK2H10
Biotin
Human pheochromocytoma
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.